Learn More
Sepax Technologies Inc Sepax Protein standard for Size Exclusion Chromatography (SEC-80 and SEC-100) and used for general correlation of elution time vs. protein MW (5 vials, 50ul/vial) -BSA (1 mg/mL) -Ribonuclease A (1 mg/mL) -Uracil (0.1 mg/mL)
SDP
Catalog No. 502336766
Encompass
Protein standards are a group of proteins that are used as the standards to calibrate various modes of separation phases listed below. Those standards are supplied as a set of 5 vials of protein mixture.
Catalog No. 50-233-6766 Supplier Sepax Technologies Inc Supplier No. 201002BRU
Protein standards are a group of proteins that are used as the standards to calibrate various modes of separation phases listed below. Those standards are supplied as a set of 5 vials of protein mixture.