Learn More
Sepax Technologies Inc Sepax Protein standard for Size Exclusion Chromatography (SEC-1000 and SEC-2000) and used for general correlation of elution time vs. protein MW (5 vials, 50ul/vial) -Thyroglobulin (1 mg/mL) -Uracil (0.1 mg/mL)
SDP
Catalog No. 502338623
Encompass
Protein standards are a group of proteins that are used as the standards to calibrate various modes of separation phases listed below. Those standards are supplied as a set of 5 vials of protein mixture.
Catalog No. 50-233-8623 Supplier Sepax Technologies Inc Supplier No. 201002TU
Protein standards are a group of proteins that are used as the standards to calibrate various modes of separation phases listed below. Those standards are supplied as a set of 5 vials of protein mixture.