Learn More
Sigma Aldrich Fine Chemicals Biosciences Sephacryl(R) 200-HR, MW range 5-250 kDa (globular proteins), MW range 1-80 kDa (dextrans) | 65546-95-4 | MFCD00165824 | 250ML
Catalog No. 501765004
Encompass
Sephacryl(R) 200-HR, MW range 5-250 kDa (globular proteins), MW range 1-80 kDa (dextrans) | 65546-95-4 | MFCD00165824 | 250ML
Catalog No. 50-176-5004 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. S200HR250ML
Sephacryl(R) 200-HR, MW range 5-250 kDa (globular proteins), MW range 1-80 kDa (dextrans) | 65546-95-4 | MFCD00165824 | 250ML