Learn More
Sigma Aldrich Fine Chemicals Biosciences Sephacryl(R) 200-HR, MW range 5-250 kDa (globular proteins), MW range 1-80 kDa (dextrans) | 65546-95-4 | MFCD00165824 | 100ML
Catalog No. 501765003
Encompass
Sephacryl(R) 200-HR, MW range 5-250 kDa (globular proteins), MW range 1-80 kDa (dextrans) | 65546-95-4 | MFCD00165824 | 100ML
Catalog No. 50-176-5003 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. S200HR100ML
Sephacryl(R) 200-HR, MW range 5-250 kDa (globular proteins), MW range 1-80 kDa (dextrans) | 65546-95-4 | MFCD00165824 | 100ML