Learn More
Sigma Aldrich Fine Chemicals Biosciences Sephacryl 300-HR, MW range 10-1500 kDa (globular proteins), MW range 1-400 kDa (dextrans) | 65546-95-4 | MFCD00165824 | 250ml
Catalog No. 501765006
Encompass
Sephacryl 300-HR, MW range 10-1500 kDa (globular proteins), MW range 1-400 kDa (dextrans) MW: | 65546-95-4 | MFCD00165824 | 250ml
Catalog No. 50-176-5006 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. S300HR250ML
Sephacryl 300-HR, MW range 10-1500 kDa (globular proteins), MW range 1-400 kDa (dextrans) MW: | 65546-95-4 | MFCD00165824 | 250ml