Learn More
Sigma Aldrich Fine Chemicals Biosciences Sephacryl(R) 300-HR, MW range 10-1500 kDa (globular proteins), MW range 1-400 kDa (dextrans) | 65546-95-4 | MFCD00165824 | 100ML
Catalog No. 501765005
Encompass
Sephacryl(R) 300-HR, MW range 10-1500 kDa (globular proteins), MW range 1-400 kDa (dextrans) | 65546-95-4 | MFCD00165824 | 100ML
Catalog No. 50-176-5005 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. S300HR100ML
Add to Cart
Add to Cart
Sephacryl(R) 300-HR, MW range 10-1500 kDa (globular proteins), MW range 1-400 kDa (dextrans) | 65546-95-4 | MFCD00165824 | 100ML