Learn More
Sigma Aldrich Fine Chemicals Biosciences Sephacryl(R) 400-HR, MW range 20-8000 kDa (globular proteins) | 65546-95-4 | MFCD00165824 | 100ML
Catalog No. 501765007
Encompass
Sephacryl(R) 400-HR, MW range 20-8000 kDa (globular proteins) | 65546-95-4 | MFCD00165824 | 100ML
Catalog No. 50-176-5007 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. S400HR100ML
Sephacryl(R) 400-HR, MW range 20-8000 kDa (globular proteins) | 65546-95-4 | MFCD00165824 | 100ML