Learn More
Sigma Aldrich Fine Chemicals Biosciences SephadexR G 10 Cytiva 17 0
Catalog No. 502904321
Encompass
Sephadex(R) G-10 is a well established gel filtration medium for desalting and buffer exchange of peptides and small biomolecules 700 molecular weight. Smallest Exlusion limit in the Sephadex(R) range.
Catalog No. 50-290-4321 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17001001
Sephadex(R) G-10 is a well established gel filtration medium for desalting and buffer exchange of peptides and small biomolecules 700 molecular weight. Smallest Exlusion limit in the Sephadex(R) range.