Learn More
Sigma Aldrich Fine Chemicals Biosciences SephadexR G 25 Superfine C
Catalog No. 502904864
Encompass
Sephadex(R) G-25 is well established gel filtration medium for desalting and buffer exchange in industrial applications.
Catalog No. 50-290-4864 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17003101
Sephadex(R) G-25 is well established gel filtration medium for desalting and buffer exchange in industrial applications.