Learn More
Sigma Aldrich Fine Chemicals Biosciences SephadexR LH-20 Cytiva 17-
Catalog No. 502891564
Encompass
Sephadex(R) LH20 is a liquid chromatography media for molecular sizing of natural product such as steroid, terpenoid, lipid, low molecular weight peptide. Useful to analytical and industrial scale for preparation of closely related molecular species.
Catalog No. 50-289-1564 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17009010
Sephadex(R) LH20 is a liquid chromatography media for molecular sizing of natural product such as steroid, terpenoid, lipid, low molecular weight peptide. Useful to analytical and industrial scale for preparation of closely related molecular species.