Learn More
Rockland Immunochemicals SEPHAROSE™ PROTEIN A - PA50-00-0005
SDP
Supplier:  Rockland Immunochemicals PA50000005
Encompass
Sepharose™ Protein A is a suspension of beads conjugated to native protein A derived from Staphylococcus aureaus. When evenly dispersed approximately 10 mL of suspension is equal to 5.0 cc of settled beads of Sepharose™ 4 Fast Flow (highly cross linked, 4% agarose derivative). The bead size is approximately 45-165 µm when swollen. Approximately 6.0 mg of Protein A is bound per cc of drained beads. Each cc of drained beads will bind approximately 35 mg Human IgG. The exclusion limit (Mr) of the beads is 3 x 10^7. The maximum linear flow rate is approximately 1300 cm/hr. This product shows long term stability within pH range of 3 to 9 and short term stability within pH range of 2 to 10.
Catalog No. 50-105-9104