Learn More
Apexbio Technology LLC Sephin1 13098-73-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061677
Encompass_Preferred
Sephin1 (CAS 13098-73-2) is a selective inhibitor of PPP1R15A a regulatory subunit of protein phosphatase 1 involved in the regulation of stress-induced eIF2 dephosphorylation Under endoplasmic reticulum (ER) stress phosphorylated eIF2 attenuates protein synthesis preventing accumulation of misfolded proteins By inhibiting PPP1R15A Sephin1 sustains eIF2 phosphorylation extending translational attenuation during ER stress In cell assays Sephin1 selectively disrupts PPP1R15A-PP1c complexes without affecting PPP1R15B-mediated complexes In mouse models harboring misfolded protein mutations (e g MPZ or SOD1) Sephin1 administration mitigates molecular cellular and functional deficits thus relevant for proteostasis-related disease studies
Catalog No. 50-000-61677 Supplier Apexbio Technology LLC Supplier No. A87085.1
Sephin1 (CAS 13098-73-2) is a selective inhibitor of PPP1R15A a regulatory subunit of protein phosphatase 1 involved in the regulation of stress-induced eIF2 dephosphorylation Under endoplasmic reticulum (ER) stress phosphorylated eIF2 attenuates protein synthesis preventing accumulation of misfolded proteins By inhibiting PPP1R15A Sephin1 sustains eIF2 phosphorylation extending translational attenuation during ER stress In cell assays Sephin1 selectively disrupts PPP1R15A-PP1c complexes without affecting PPP1R15B-mediated complexes In mouse models harboring misfolded protein mutations (e g MPZ or SOD1) Sephin1 administration mitigates molecular cellular and functional deficits thus relevant for proteostasis-related disease studies