Learn More
Aviva Systems Biology Corp SEPHS1 Antibody - C-terminal region (ARP48737_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48737_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SEPHS1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SEPHS1. Alternate names: MGC4980, SELD, SPS, SPS1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59907