Learn More
Medchemexpress LLC Sepimostat (FUT-187 free base) | 103926-64-3 | MFCD00868988 | 98.0% | 373.4 g/mol | C21H19N5O2 | 5 MG
SDP
Catalog No. 5000221830
Encompass_Preferred
Sepimostat (FUT-187 free base) is a research compound reported to act as a selective NR2B-subunit N-methyl-D-aspartate (NMDA) receptor antagonist. It has shown neuroprotective activity in preclinical studies and inhibits Ifenprodil binding with a reported Ki of 27.7 μM. For research use only.
  • Selective NR2B NMDA receptor antagonist suitable for mechanism-of-action studies.
  • Demonstrated neuroprotective activity in preclinical models.
  • Inhibits Ifenprodil binding (reported Ki ≈ 27.7 μM).
  • Available as a solid and as a DMSO solution for in vitro assays.
  • Supplied in multiple milligram quantities to support assay development and screening.
Catalog No. 50-002-21830 Supplier Medchemexpress LLC Supplier No. HY1362995MG
Sepimostat (FUT-187 free base) is a research compound reported to act as a selective NR2B-subunit N-methyl-D-aspartate (NMDA) receptor antagonist. It has shown neuroprotective activity in preclinical studies and inhibits Ifenprodil binding with a reported Ki of 27.7 μM. For research use only.
  • Selective NR2B NMDA receptor antagonist suitable for mechanism-of-action studies.
  • Demonstrated neuroprotective activity in preclinical models.
  • Inhibits Ifenprodil binding (reported Ki ≈ 27.7 μM).
  • Available as a solid and as a DMSO solution for in vitro assays.
  • Supplied in multiple milligram quantities to support assay development and screening.