Learn More
STEMCELL Technologies SepMate™-15 (IVD)
SDP
Catalog No. NC0708026
Encompass
Simplify peripheral blood mononuclear cell (PBMC) isolation by incorporating SepMate™ into your density gradient centrifugation step. SepMate™ tubes contain an insert that creates a barrier between the density gradient medium and blood, thus eliminating the need for careful blood layering and allowing mononuclear cells to be easily harvested with a simple pour. This product can be used with RosetteSep™ to isolate specific immune cell subsets.SepMate™ is manufactured under cGMP and registered as an In Vitro Diagnostic device in Australia, Canada, Europe, and the USA. In China, SepMate™ is considered a nonmedical device by the China Food and Drug Administration and should be used as general lab equipment. The end user is responsible for determining whether the product is suitable for their specific application.Browse our Frequently Asked Questions (FAQs) (https://www.stemcell.com/sepmate-faq.html) on SepMate™.
Catalog No. NC0708026 Supplier STEMCELL Technologies Supplier No. 85415
Simplify peripheral blood mononuclear cell (PBMC) isolation by incorporating SepMate™ into your density gradient centrifugation step. SepMate™ tubes contain an insert that creates a barrier between the density gradient medium and blood, thus eliminating the need for careful blood layering and allowing mononuclear cells to be easily harvested with a simple pour. This product can be used with RosetteSep™ to isolate specific immune cell subsets.SepMate™ is manufactured under cGMP and registered as an In Vitro Diagnostic device in Australia, Canada, Europe, and the USA. In China, SepMate™ is considered a nonmedical device by the China Food and Drug Administration and should be used as general lab equipment. The end user is responsible for determining whether the product is suitable for their specific application.Browse our Frequently Asked Questions (FAQs) (https://www.stemcell.com/sepmate-faq.html) on SepMate™.