Learn More
Aviva Systems Biology Corp SELENON Antibody - C-terminal region (ARP47655_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47655_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SEPN1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SEPN1. Alternate names: FLJ24021, MDRS1, RSMD1, RSS, SELN, CFTDApplicationWB, IHC. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-53524