Learn More
Waters Corp Sep-Pak 96-well μElution Plate, tC18, 5 mg Sorbent per Well, 37–55 μm
Catalog No. 50818653 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Sep-Pak tC18 is a silica-based bonded phase featuring strong hydrophobicity and trifunctional bonding chemistry, which provides increased hydrolytic stability compared to conventional C18 bonded phases. The innovative 96-well plate design ensures compatibility with most liquid handling and SPE automation systems, making it an ideal complement for high-throughput laboratory applications.
  • tC18 sorbent type with strong hydrophobicity.
  • 96-well μElution plate format.
  • 5 mg sorbent per well.
  • 37 to 55 μm particle size.
  • 17 % carbon load.
  • Endcapped.
  • Mass spec compatible.
  • Reversed phase mode.
  • 125 Å pore size.
  • Silica sorbent substrate.
  • Not water wettable.
  • pH range of 2 to 8.
  • Polypropylene material.
Catalog No. 50-818-653 Supplier Waters Corp Supplier No. 186002318
Sep-Pak tC18 is a silica-based bonded phase featuring strong hydrophobicity and trifunctional bonding chemistry, which provides increased hydrolytic stability compared to conventional C18 bonded phases. The innovative 96-well plate design ensures compatibility with most liquid handling and SPE automation systems, making it an ideal complement for high-throughput laboratory applications.
  • tC18 sorbent type with strong hydrophobicity.
  • 96-well μElution plate format.
  • 5 mg sorbent per well.
  • 37 to 55 μm particle size.
  • 17 % carbon load.
  • Endcapped.
  • Mass spec compatible.
  • Reversed phase mode.
  • 125 Å pore size.
  • Silica sorbent substrate.
  • Not water wettable.
  • pH range of 2 to 8.
  • Polypropylene material.