Learn More
Waters Corp Sep-Pak C18 Vac Cartridge, 3 cc, 500 mg, 55 to 105 um
Catalog No. 50818603 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This C18 3 cc Vac Cartridge features a silica-based octadecyl bonded phase with strong hydrophobicity, suitable for adsorbing weakly hydrophobic analytes from aqueous solutions. It is designed to simplify liquid chromatography protocols, enhance safety, and facilitate direct field sampling.
  • Adsorbs weakly hydrophobic analytes from aqueous solutions
  • Used for solvent exchange and removal of post-analyte constituents in liquid chromatography
  • Sorbent per cartridge: 500 mg
  • Particle size: 55 to 105 μm
  • Units per package: 50 pk
  • pH range: 2 to 8
  • Barrel size: 3 cc
  • Mass Spec compatible
Catalog No. 50-818-603 Supplier Waters Corp Supplier No. WAT020805
This C18 3 cc Vac Cartridge features a silica-based octadecyl bonded phase with strong hydrophobicity, suitable for adsorbing weakly hydrophobic analytes from aqueous solutions. It is designed to simplify liquid chromatography protocols, enhance safety, and facilitate direct field sampling.
  • Adsorbs weakly hydrophobic analytes from aqueous solutions
  • Used for solvent exchange and removal of post-analyte constituents in liquid chromatography
  • Sorbent per cartridge: 500 mg
  • Particle size: 55 to 105 μm
  • Units per package: 50 pk
  • pH range: 2 to 8
  • Barrel size: 3 cc
  • Mass Spec compatible