Learn More
Waters Corp Waters Sep-Pak tC18 Vac Cartridge, 1 cc, 50 mg Sorbent
Catalog No. 50818644 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Sep-Pak tC18 Vac Cartridge is a solid phase extraction (SPE) product designed for sample preparation. It features a strongly hydrophobic, silica-based bonded phase with trifunctional bonding chemistry, offering enhanced hydrolytic stability.
  • Strongly hydrophobic, silica-based bonded phase.
  • Trifunctional bonding chemistry for enhanced hydrolytic stability.
  • Reduces solvent consumption and increases recoveries.
  • Optimal surface area, pore size, and particle size distributions.
  • Packing weight controlled within ±5% for reproducible performance.
  • Sealed in a non-contaminating poly-foil pouch.
  • Molded polypropylene syringe-barrel bodies with polyethylene frits.
  • Standard male-Luer tips for multi-position vacuum manifolds.
  • Manufactured in ISO 13485/9002 compliant facilities.
Catalog No. 50-818-644 Supplier Waters Corp Supplier No. WAT054960
The Sep-Pak tC18 Vac Cartridge is a solid phase extraction (SPE) product designed for sample preparation. It features a strongly hydrophobic, silica-based bonded phase with trifunctional bonding chemistry, offering enhanced hydrolytic stability.
  • Strongly hydrophobic, silica-based bonded phase.
  • Trifunctional bonding chemistry for enhanced hydrolytic stability.
  • Reduces solvent consumption and increases recoveries.
  • Optimal surface area, pore size, and particle size distributions.
  • Packing weight controlled within ±5% for reproducible performance.
  • Sealed in a non-contaminating poly-foil pouch.
  • Molded polypropylene syringe-barrel bodies with polyethylene frits.
  • Standard male-Luer tips for multi-position vacuum manifolds.
  • Manufactured in ISO 13485/9002 compliant facilities.