Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc Sepra C18-E (50 µm, 65 Å)

Catalog No. 502393978 Shop All Phenomenex Products
Encompass

Sepra C18-E (50 µm, 65 Å) Bulk Packing, 3 kg

Catalog No. 50-239-3978 Supplier Phenomenex Inc Supplier No. 04V4348
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Sepra C18-E (50 µm, 65 Å) Bulk Packing, 3 kg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.