Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc Bulk Packing, 100 g

Supplier:  Phenomenex Inc 04G4607

Encompass

Sepra™ ZTL-WCX (100 m, 300 ) 04G-4607

Catalog No. 50-239-5323


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.