Learn More
Medchemexpress LLC Seproxetine hydrochloride | 127685-30-7 | MFCD06658520 | 93.1% | 331.76 g·mol⁻1 | C16H17ClF3NO | 100MG
SDP
Catalog No. 5000235571
Encompass_Preferred
Seproxetine hydrochloride is the hydrochloride salt of seproxetine (S-norfluoxetine), a selective serotonin reuptake inhibitor (SSRI) and an active metabolite of fluoxetine. Intended for research use in biochemical and pharmacological studies, the compound is provided in multiple small-scale package sizes and is characterized by its molecular formula C16H17ClF3NO, molecular weight 331.76 g·mol⁻1, and CAS 127685-30-7.
  • High enantiomeric excess and purity for research applications (97.6% ee; 93.1% purity).
  • Available in multiple small package sizes to support screening and synthesis.
  • Provided as the hydrochloride salt for improved handling and stability.
  • Documented molecular formula and weight for accurate formulation and calculations.
  • CAS number included for unambiguous chemical identification.
Catalog No. 50-002-35571 Supplier Medchemexpress LLC Supplier No. HY124357100MG
Seproxetine hydrochloride is the hydrochloride salt of seproxetine (S-norfluoxetine), a selective serotonin reuptake inhibitor (SSRI) and an active metabolite of fluoxetine. Intended for research use in biochemical and pharmacological studies, the compound is provided in multiple small-scale package sizes and is characterized by its molecular formula C16H17ClF3NO, molecular weight 331.76 g·mol⁻1, and CAS 127685-30-7.
  • High enantiomeric excess and purity for research applications (97.6% ee; 93.1% purity).
  • Available in multiple small package sizes to support screening and synthesis.
  • Provided as the hydrochloride salt for improved handling and stability.
  • Documented molecular formula and weight for accurate formulation and calculations.
  • CAS number included for unambiguous chemical identification.