Learn More
Waters Corp Waters Check Valve Cartridge, Ceramic, 2-Pack, for Alliance HPLC System
Catalog No. 50818922 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This check valve cartridge is a replacement component designed for specific separations modules within HPLC systems. It features a durable ceramic ball and seat, offering an optional solution for addressing sticking issues. Each package contains two check valves, ensuring convenient replacement for maintaining optimal instrument performance.
  • Replacement check valve cartridge.
  • Features a ceramic ball and seat.
  • Optional check valve to help with sticking issues.
  • Supplied as a 2-pack.
Catalog No. 50-818-922 Supplier Waters Corp Supplier No. 700002333
This check valve cartridge is a replacement component designed for specific separations modules within HPLC systems. It features a durable ceramic ball and seat, offering an optional solution for addressing sticking issues. Each package contains two check valves, ensuring convenient replacement for maintaining optimal instrument performance.
  • Replacement check valve cartridge.
  • Features a ceramic ball and seat.
  • Optional check valve to help with sticking issues.
  • Supplied as a 2-pack.