Learn More
Aviva Systems Biology Corp SEPT12 Antibody - middle region (ARP52696_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52696_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Sept12. It was validated on Western Blot. . Alternate names: 1700028G04Rik, 4933413B09Rik, MGC144518, Septin12ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-285-757