Learn More
Aviva Systems Biology Corp SEPT4 Antibody - N-terminal region (AVARP02014_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP02014_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SEPT4 (septin-4). It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human SEPT4 (septin-4). Alternate names: H5, ARTS, MART, SEP4, CE5B3, PNUTL2, hucep-7, BRADEION, hCDCREL-2ApplicationIHC, WB. Species reactivity: Dog, Human
Catalog No. 50-014-53771