Learn More
Trajan Scientific SEPTA 11MM EC 25/PK
SDP
Catalog No. 502530246
Encompass_Preferred
Trajan Septa for Agilent, Diameter Outer: 11 mm, Temperature Max: 350 deg.C, Long Life EC Septa, Description: Combines significantly longer injection life, low bleed and low injection portadhesion., High Temperature Silicone, Quantity: 25/Pk.
Catalog No. 50-253-0246 Supplier Trajan Scientific Supplier No. 041902
Trajan Septa for Agilent, Diameter Outer: 11 mm, Temperature Max: 350 deg.C, Long Life EC Septa, Description: Combines significantly longer injection life, low bleed and low injection portadhesion., High Temperature Silicone, Quantity: 25/Pk.