Learn More
MilliporeSigma™ Supelco™ Septa for 12 x 32 mm Standard Screw-Thread Autosampler Vials
Catalog No. 111012015 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Color:
Red, White
White
Material:
PTFE
PTFE/Silicone
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Material
11-101-2015 Red, White PTFE/Silicone
11-101-1337 White PTFE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-2015 Supplier MilliporeSigma™ Supelco™ Supplier No. Z2916761PAK
Add to Cart
Add to Cart
It is used as an air-tight seal, and prevents contamination
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Inner 8 mm
Color Red, White
Material PTFE/Silicone
No. per Pack 100
Thickness (English) 0.065 in.