Learn More
Ace Glass, Inc. Septa, white rubber, sized for 13-18mm OD tubes, sp/12
SDP
Catalog No. 50976682
Encompass
Septa, white rubber, sized for 13-18mm OD tubes, sp/12
Catalog No. 50-976-682 Supplier Ace Glass, Inc. Supplier No. 909649
Septa, white rubber, sized for 13-18mm OD tubes, sp/12