Learn More
Microsolv Technology Corporation Septa, 13mm, PTFE Disc for use in 13-425mm thread, 15x45mm autosampler vial caps. Caps sold separately. 100/PK
Catalog No. 502271350
Encompass_Preferred
This PTFE (only) Septa is for single injections or non-volatile samples as it will not reseal. This Septa is used in Caps for 4ml vials.
This item is not returnable. View return policy
Catalog No. 50-227-1350 Supplier Microsolv Technology Corporation Supplier No. 9504S05PTFE
This PTFE (only) Septa is for single injections or non-volatile samples as it will not reseal. This Septa is used in Caps for 4ml vials.