Learn More
Microsolv Technology Corporation Septa, 17.5mm, Headspace, Reddish Brown Butyl Rubber / Grey PTFE. For use in 18mm Aluminum, Screw Caps, sold separately. 1000/CS.
Catalog No. 502271209
Encompass_Preferred
Butyl Rubber / PTFE Septa, Durometer of 55 Shore A (hardness) and slightly thicker 1.6mm compared with other Septa for 18mm Screw Caps makes these Caps useful as Septa or Liners with ND18, Precision Thread Screw Caps for Headspace. Supplied in a polyethylene bag.
This item is not returnable. View return policy
Catalog No. 50-227-1209 Supplier Microsolv Technology Corporation Supplier No. 9502521
Butyl Rubber / PTFE Septa, Durometer of 55 Shore A (hardness) and slightly thicker 1.6mm compared with other Septa for 18mm Screw Caps makes these Caps useful as Septa or Liners with ND18, Precision Thread Screw Caps for Headspace. Supplied in a polyethylene bag.