Learn More
Microsolv Technology Corporation Septa, 17.5mm, Headspace, Transparent, Bluish Silicone Rubber / White PTFE. For use in 18mm Aluminum, Screw Caps, sold separately. 1000/CS.
Catalog No. 502271210
Encompass_Preferred
Silicone Rubber / Ultra Clean PTFE, with Durometer of 45 Shore A, (hardness) and 1.3mm thick is good for sensitive analyses. The lower Durometer means that this is a softer Septa and therefore better for easier penetrations by autosampler needles. Use in ND18, Precision Thread Screw Caps for Headspace autosamplers. Supplied in Polyethylene Bags.
This item is not returnable. View return policy
Catalog No. 50-227-1210 Supplier Microsolv Technology Corporation Supplier No. 9502522
Silicone Rubber / Ultra Clean PTFE, with Durometer of 45 Shore A, (hardness) and 1.3mm thick is good for sensitive analyses. The lower Durometer means that this is a softer Septa and therefore better for easier penetrations by autosampler needles. Use in ND18, Precision Thread Screw Caps for Headspace autosamplers. Supplied in Polyethylene Bags.