Learn More
Microsolv Technology Corporation Septa, 17.5mm, Headspace, White Silicone Rubber / Blue PTFE. For use in 18mm Aluminum, Screw Caps, sold separately. 1000/CS.
Supplier:  Microsolv Technology Corporation 9502023
Encompass_Preferred
Silicone Rubber / PTFE Septa, Durometer of 55 Shore A (hardness) and 1.5mm thick. Temperature resistant from -60°C up to 200°C. Use in ND18, Precision Thread Screw Caps for Headspace. Supplied in a polyethylene bag.
Catalog No. 50-227-1190