Learn More
Microsolv Technology Corporation Caps, Headspace, Crimp Top, with Dark Grey Butyl Rubber Septa, fitted in Steel, Magnetic, Gold Caps with 5mm center holes. For use with 20mm Crimp Top Headspace Vials. 100/PK.
Supplier:  Microsolv Technology Corporation 95025013D
Encompass_Preferred
Molded, Butyl Rubber Septa is 55⁰ Shore A (hardness) and is 3.0mm thick. Temperature resistant from -40C to 120C. Recommended for non critical analyses or when PTFE is an analyte or must be avoided. Recommended for CTC™ and other autosamplers. Low cost for budget minded laboratories.
Catalog No. 50-227-1198
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.