Learn More
Microsolv Technology Corporation Septa, 20mm, Headspace, Molded, Grey Butyl Rubber / Light Grey Rubber with Laminated PTFE. For use in 20mm Crimp Caps, sold separately. Pharma-Fix type Septa. 1000/CS.
Catalog No. 502271201
Encompass_Preferred
Shaped, Butyl Rubber with Durometer of 50 Shore A, (hardness) and 3.0mm thick. Partially laminated with PTFE only on the light grey side of the Septa that faces the sample. Butyl Rubber facing the glass vial provides a very good seal, due to its elasticity. Combining inertness of PTFE and elasticity of Butyl Rubber. This type of Septa is commonly known as Pharma-Fix™ and is temperature resistant from -40⁰ to 120⁰C. Use in Aluminum or Steel Crimp Caps for Headspace autosamplers. Supplied in Polyethylene Bags.
This item is not returnable. View return policy
Catalog No. 50-227-1201 Supplier Microsolv Technology Corporation Supplier No. 9502503
Shaped, Butyl Rubber with Durometer of 50 Shore A, (hardness) and 3.0mm thick. Partially laminated with PTFE only on the light grey side of the Septa that faces the sample. Butyl Rubber facing the glass vial provides a very good seal, due to its elasticity. Combining inertness of PTFE and elasticity of Butyl Rubber. This type of Septa is commonly known as Pharma-Fix™ and is temperature resistant from -40⁰ to 120⁰C. Use in Aluminum or Steel Crimp Caps for Headspace autosamplers. Supplied in Polyethylene Bags.