Learn More
Microsolv Technology Corporation Septa, 20mm, Headspace, Molded Butyl Rubber, Grey, completely laminated with transparent PTFE. For use in 20mm Crimp Caps, sold separately. 1000/CS.
Catalog No. 502271200
Encompass_Preferred
Shaped, Butyl Rubber with Durometer of 50 Shore A, (hardness) and 3.0mm thick. Completely laminated with PTFE on both sides of the Septa. Temperature resistant from -40⁰ to 120⁰C. Use in Aluminum or Steel Crimp Caps for Headspace autosamplers. Supplied in Polyethylene Bags. Can also be used as a stopper.
This item is not returnable. View return policy
Catalog No. 50-227-1200 Supplier Microsolv Technology Corporation Supplier No. 9502502
Shaped, Butyl Rubber with Durometer of 50 Shore A, (hardness) and 3.0mm thick. Completely laminated with PTFE on both sides of the Septa. Temperature resistant from -40⁰ to 120⁰C. Use in Aluminum or Steel Crimp Caps for Headspace autosamplers. Supplied in Polyethylene Bags. Can also be used as a stopper.