Learn More
Microsolv Technology Corporation Septa 20Mm Hs Molded 100/Pk
Catalog No. NC1925980
Encompass_Preferred
Septa, 20mm, Headspace, Molded Butyl Rubber, Grey, Stopper. For use in 20mm Crimp Caps, sold separately. MicroSolv Brand. 100/PK
Catalog No. NC1925980 Supplier Microsolv Technology Corporation Supplier No. 9502506
Septa, 20mm, Headspace, Molded Butyl Rubber, Grey, Stopper. For use in 20mm Crimp Caps, sold separately. MicroSolv Brand. 100/PK