Learn More
Microsolv Technology Corporation Septa, 20mm, Headspace, White Silicone Rubber / Blue PTFE Foil. For use in 20mm Crimp Caps, sold separately. 1000/CS.
Catalog No. 502269900
Encompass_Preferred
Silicone Rubber / PTFE Septa with Durometer of 50 Shore A, (hardness) and 3.0mm thick is good for higher temperature analyses. Temperature range from -60⁰ to 200⁰C. Use in Aluminum or Steel Crimp Caps for Headspace autosamplers. Supplied in Polyethylene Bags.
This item is not returnable. View return policy
Catalog No. 50-226-9900 Supplier Microsolv Technology Corporation Supplier No. 9502523
Silicone Rubber / PTFE Septa with Durometer of 50 Shore A, (hardness) and 3.0mm thick is good for higher temperature analyses. Temperature range from -60⁰ to 200⁰C. Use in Aluminum or Steel Crimp Caps for Headspace autosamplers. Supplied in Polyethylene Bags.