Learn More
Microsolv Technology Corporation Septa, 20mm, Headspace, Blue Silicone Rubber completely laminated with Transparent PTFE. For use in 20mm Crimp Caps, sold separately. 1000/CS.
Catalog No. 502271205
Encompass_Preferred
Silicone Rubber with Durometer of 45 Shore A, (hardness) and 3.0mm thick. Completely laminated with Ultra Clean PTFE on both sides of the Septa which is good for sensitive analyses. The lower Durometer means that this is a softer Septa and therefore better for easier penetrations by autosampler needles. Wide temperature range from -60⁰ to 200⁰C. Use in Aluminum or Steel Crimp Caps for Headspace autosamplers. Supplied in Polyethylene Bags.
This item is not returnable. View return policy
Catalog No. 50-227-1205 Supplier Microsolv Technology Corporation Supplier No. 9502504
Silicone Rubber with Durometer of 45 Shore A, (hardness) and 3.0mm thick. Completely laminated with Ultra Clean PTFE on both sides of the Septa which is good for sensitive analyses. The lower Durometer means that this is a softer Septa and therefore better for easier penetrations by autosampler needles. Wide temperature range from -60⁰ to 200⁰C. Use in Aluminum or Steel Crimp Caps for Headspace autosamplers. Supplied in Polyethylene Bags.