Learn More
Microsolv Technology Corporation Septa, 20mm, Headspace, White Silicone Rubber / White PTFE. For use in 20mm Crimp Caps, sold separately. Ultra Low Bleed. 1000/CS.
Catalog No. 502270352
Encompass_Preferred
Silicone Rubber / PTFE Septa with Durometer of 45 Shore A, (hardness) and 3.0mm thick with Ultra Clean PTFE and Ultra Low Bleed Rubber is good for highly sensitive analyses. The lower Durometer means that this is a softer Septa and therefore better for easier penetrations by autosampler needles. Wide temperature range from -60⁰ to 200⁰C. Use in Aluminum or Steel Crimp Caps for Headspace autosamplers. Supplied in Polyethylene Bags.
Catalog No. 50-227-0352 Supplier Microsolv Technology Corporation Supplier No. 9502508
Silicone Rubber / PTFE Septa with Durometer of 45 Shore A, (hardness) and 3.0mm thick with Ultra Clean PTFE and Ultra Low Bleed Rubber is good for highly sensitive analyses. The lower Durometer means that this is a softer Septa and therefore better for easier penetrations by autosampler needles. Wide temperature range from -60⁰ to 200⁰C. Use in Aluminum or Steel Crimp Caps for Headspace autosamplers. Supplied in Polyethylene Bags.