Learn More
Microsolv Technology Corporation Septa, 20mm, Headspace, White Silicone Rubber / Silver Foil. For use in 20mm Crimp Caps, sold separately. High Temperature. (10x100) 1000/CS.
Catalog No. 502271207
Encompass_Preferred
Silicone Rubber / Aluminum Foil Silver Septa with Durometer of 50 Shore A, (hardness) and 3.0mm thick is good for high temperature analyses. No color has been added to this Septa. Often used with Perkin Elmer Instruments.Wide temperature range from -60⁰ to 220⁰C. Use in Aluminum or Steel Crimp Caps for Headspace autosamplers. Supplied in Polyethylene Bags.
This item is not returnable. View return policy
Catalog No. 50-227-1207 Supplier Microsolv Technology Corporation Supplier No. 9502514
Silicone Rubber / Aluminum Foil Silver Septa with Durometer of 50 Shore A, (hardness) and 3.0mm thick is good for high temperature analyses. No color has been added to this Septa. Often used with Perkin Elmer Instruments.Wide temperature range from -60⁰ to 220⁰C. Use in Aluminum or Steel Crimp Caps for Headspace autosamplers. Supplied in Polyethylene Bags.