Learn More
Waters Corp 8mm PTFE Septum, 100/pk
Supplier:  Waters Corp WAT058886
Encompass_Preferred
Waters offers a selection of cap, septum, and assembled cap/septum for various vial sizes. The range of septum material is limited to PTFE, silicone / PTFE, and polyethylene septumless caps.
Catalog No. 50-785-845