Learn More
Microsolv Technology Corporation Septa, 8mm, White Silicone Rubber / Beige PTFE that is 0.60" thick and 0.319" diameter for use in 8-425mm thread caps. Caps sold separately. 1000/CS.
Catalog No. 502270511
Encompass_Preferred
Used for both HPLC or GC, 8mm screw caps.
This item is not returnable. View return policy
Catalog No. 50-227-0511 Supplier Microsolv Technology Corporation Supplier No. 96008105010
Used for both HPLC or GC, 8mm screw caps.