Learn More
Microsolv Technology Corporation Septa, 9mm, Orange Silicone Rubber / White PTFE that is 0.40" thick and 0.344" diameter for use in 9-425mm thread caps. Caps sold separately. 1000/CS.
Catalog No. 502270512
Encompass_Preferred
This Septa is packaged without the lubricant, Talc and some of the Septa may stick together during shipping, but they will not introduce extractables to the chromatography. This Septa is cut for use in Agilent style screw caps.
This item is not returnable. View return policy
Catalog No. 50-227-0512 Supplier Microsolv Technology Corporation Supplier No. 9600910T4010
This Septa is packaged without the lubricant, Talc and some of the Septa may stick together during shipping, but they will not introduce extractables to the chromatography. This Septa is cut for use in Agilent style screw caps.