Learn More
Mocon Inc SEPTA ADHESIVE 1/SIDE 1000/PK
SDP
Catalog No. NC0610836
Encompass
Pack of septa. Used to form a seal when the needle in inserted into a package to test the headspace contents 1000 septa per pack.
Catalog No. NC0610836 Supplier Mocon Inc Supplier No. 940301
Pack of septa. Used to form a seal when the needle in inserted into a package to test the headspace contents 1000 septa per pack.