Learn More
C&G Containers SEPTA CAP BOND 24/400 PTFE/SIL
SDP
Catalog No. NC9855154
Encompass_Preferred
24-414 Wht PP 1piece Hole Cap Low Bleed Inter-seal Bonded w/125mil Silicone/PTFE Septa(Natural) 144/pk
Catalog No. NC9855154 Supplier C&G Containers Supplier No. CAH2420050
24-414 Wht PP 1piece Hole Cap Low Bleed Inter-seal Bonded w/125mil Silicone/PTFE Septa(Natural) 144/pk