Learn More
Microsolv Technology Corporation Septa For 8Mm Caps Sil/Ptfese
Catalog No. 5001452407
Encompass_Preferred
Septa, 8mm, White Silicone Rubber / White PTFE that is 0.45"" thick and 0.321"" diameter for use in 8-425mm thread caps. Caps sold separately. MicroSolv Brand. 1000/CS.
Catalog No. 50-014-52407 Supplier Microsolv Technology Corporation Supplier No. 9582S10XK
Septa, 8mm, White Silicone Rubber / White PTFE that is 0.45"" thick and 0.321"" diameter for use in 8-425mm thread caps. Caps sold separately. MicroSolv Brand. 1000/CS.