Learn More
Trajan Scientific Septa Inlet AG3 Low Bleed 25/PK
SDP
Catalog No. 5000494043
Encompass_Preferred
6 4 mm (1/4 ) OD AG3 low-bleed GC septa PK25
Catalog No. 50-004-94043 Supplier Trajan Scientific Supplier No. 041916
6 4 mm (1/4 ) OD AG3 low-bleed GC septa PK25