Learn More
Trajan Scientific Septa Inlet AG3 Low Bleed 50/PK
SDP
Catalog No. 5000493559
Encompass_Preferred
11 mm OD AG3 low-bleed center guide GC septa PK50
This item is not returnable. View return policy
Catalog No. 50-004-93559 Supplier Trajan Scientific Supplier No. 246225
11 mm OD AG3 low-bleed center guide GC septa PK50