Learn More
Trajan Scientific Septa Inlet BTO Premium 25/PK
SDP
Catalog No. 5000494752
Encompass_Preferred
9 5 mm (3/8 ) OD BTO premium high-temp GC septa PK25
Catalog No. 50-004-94752 Supplier Trajan Scientific Supplier No. 041897
9 5 mm (3/8 ) OD BTO premium high-temp GC septa PK25