Learn More
Trajan Scientific Septa Inlet BTO Premium 50/PK
SDP
Supplier:  Trajan Scientific 298735
Encompass_Preferred
Shimadzu Plug BTO premium high-temp center guide GC septa PK50
Catalog No. 50-004-94221